Would you like to log in again?
Centrifuge Vortex Mixer, CVP-2, Grant Instruments
Supplier: Grant
CVP-2USA
10809-106EA 3400.59 USD InStock
10809-106
Centrifuge Vortex Mixer, CVP-2, Grant Instruments
Shakers and Mixers Vortex Mixers
The CVP-2 is an all-in-one PCR centrifuge/vortex mixer that allows for the simultaneous sample preparation of multiple samples at one time
The CVP-2 offers the versatility of three devices in one: centrifuge with vortex mixing, PCR plate centrifuge, and PCR plate mixer.
Independent vortex and centrifuge timers allow up to 999 cycles. Users can consistently prepare up to 192 samples simultaneously.
This versatile instrument is able to hold un-skirted, semi-skirted, and fully-skirted PCR plates with no additional accessories required.
Cycle rate: 1 to 999
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty