Would you like to log in again?
UXT Xtra Tough Heavy Pad, Brady
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
107849
10809-774RL 377.78 USD InStock
10809-774
UXT Xtra Tough Heavy Pad, Brady
Spill Control Materials
Features high performance, this all-purpose sorbent is made to handle virtually any liquid (water, petroleum and chemical-based).3-ply construction; double-sided, low-linting coverstock and meltblown core; heavy weight perfed pads
Premium extra-tough universal sorbent for industrial maintenance and hazmat, abrasion resistant and non-abrasive performance, perfect for wipe down applications. Extra strength allows wringing and re-using of sorbents. Polypropylene material. Gray color.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty