Would you like to log in again?
MRO Plus™ Heavy Pad, Brady
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
107720
10809-776CS 245.97 USD InStock
10809-776
MRO Plus™ Heavy Pad, Brady
Spill Control Materials
The MRO Plus™ Heavy Pad absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
3-ply construction makes it stronger, more durable and reduces lint. Gray color masks drips and spills, dimpled, perfed so you can take only what you need and reduce waste, full sized, heavy weight pad. For use with oil and water-based fluids. Polypropylene material. Gray color.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty