Would you like to log in again?
Vapor Pad, Cancer Diagnostics
Supplier: Cancer Diagnostics
VP01114
10809-806PK 409.29 USD InStock
10809-806
Vapor Pad, Cancer Diagnostics
Spill Control Materials
Formalin Neutralizer Pads encapsulate and neutralize harmful vapors to greatly reduce exposure to Formaldehyde.
  • Offers contrast for all biopsies
  • Cut resistant
  • Durable
  • Disposable
  • Neutralizes harmful vapors
Formalin Neutralizer Pads that encapsulate and neutralize harmful vapors to greatly reduce exposure to Formaldehyde. Light blue contained work surface offers contrast for all biopsies. Cut resistant, durable and disposable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty