Would you like to log in again?
Tissue Marking Dye, Flip-Top, Cancer Diagnostics
Supplier: Cancer Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0723-6 0723-7 0728-9 0723-9 0728-8 0728-1 0728-6 0727-2 0727-4 0723-2 0728-5 0727-9 0723-5 0728-3 0727-5 0728-7 0723-4 0723-1 0723-8 0727-8 0727-10 0728-2 0728-4 0728-10 0727-7 0727-6 0727-1 0723-3 0727-3 0723-10
10809-912EA 237.8 USD InStock
10809-912 10809-856 10809-812 10809-854 10809-852 10809-830 10809-850 10809-918 10809-818 10809-916 10809-816 10809-914 10809-814 10809-858 10809-824 10809-846 10809-922 10809-822 10809-920 10809-820 10809-864 10809-862 10809-860 10810-002 10809-828 10809-926 10809-826 10809-848 10810-000 10809-924
Tissue Marking Dye, Flip-Top, Cancer Diagnostics
Stains Histology Stains
Tissue marking dyes are used for marking margins & multiple biopsies.
  • Margins defined without bleeding
  • Dyes won't change color or fade
  • Flip-top dispenser cap
Tissue marking dyes for marking margins & multiple biopsies. Dyes have been developed to define margins without bleeding, changing color or fading. Flip-top dispenser cap.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty