Easy access to products and protocols for research use only in the identification of 2019-nCoV based on Centers for Disease Control and Prevention (CDC) recommendations
So much has changed during this unprecedented time, except your ability to count on Avantor. We continue to set science in motion to create a better world by providing you with the right solutions to keep moving forward.
Our solutions, developed with you as our focus, are crafted by our team and network of professionals with advanced degrees in science, quality control, engineering, manufacturing and industry experience.
Avantor supports end-to-end fluid management solutions – including peristaltic pumps and aseptic fluid transfer solutions – that are reliable and customer-centric, helping bioprocessing manufacturers meet their research and production goals.
A strong, vibrant research and development group is the lifeblood of all industries. VWR will support you from the latest life science products to the guaranteed purity of organic building blocks...
VWR is ready to support your production facility with reliable access to raw materials and essential supplies. We can also help you increase productivity...
VWR is proud of our years of experience providing choice and excellent service to the Industrial market from Food & Beverage, Petrochemical, Environmental Testing, Waste Water, Cosmetics, Consumer Goods, Agriculture and more...
VWR is your complete source for workplace supplies. Binders, calendars, pens, cleaning and sanitation supplies, and office equipment are just some of the essential products we offer...
New Avantor® J.T.Baker® premium conductive and non-conductive robotic tips deliver superior quality and reliable performance for results you can trust.
Avantor Services provides a wide range of specialized services and digital solutions to help you solve complex challenges.
We’ve built our reputation on consistent, comprehensive mastery of day-to-day operations, allowing lab, clinical, and production environments to focus their high-value resources on core scientific priorities.
As our customers’ needs have evolved, so have our capabilities. We have become experts in scientific operations, improving performance with sophisticated solutions and providing guidance on best practices.
You can select and customize services for peak efficiency, quality, and accelerated innovation.
The Equipment & Instrument Service Team’s focus is to provide comprehensive service solutions for our customers. We provide validation, calibration, preventative maintenance, and extended warranties on all equipment & instruments in and around the laboratory.
Synonyms:Tetrabromofluorescein disodium salt
, Eosin Y disodium salt
, Eosin 225
, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt
, Solvent Red 43
, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate
, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
, Field's Stain B
, 2,4,5,7-Tetrabromofluorescein disodium salt
ETV4
10809-902CS249.33USDInStock
10809-902
Eosin Y (yellowish) solution Type V stain for histology
Eosin Y (yellowish)
Eosin Type V is a class formulation of the alcoholic eosin-Y counterstain.
Eosin Type V produces brilliant counterstaining and differentiation of cell cytoplasms and other non-nuclear tissue components. Eosin Type V is ready to use and has been balanced to the proper PH for consistency and stability.
New Product
Formula: C₂₀H₆Br₄Na₂O₅ MW: 691.86 g/mol
MDL Number: MFCD00005040 CAS Number: 17372-87-1 UN: 1170 ADR: 3,III