Would you like to log in again?
Blueing reagent working solution, Pureview PH-Blue (pH 8)
Supplier: Cancer Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pureview
16702-4
10809-908CS 447.28 USD InStock
10809-908
Blueing reagent working solution, Pureview PH-Blue (pH 8)
Stains Microscopy and staining reagents
The Endure Staining System is designed to produce precise, consistent H&E staining.
A bluing reagent buffered to a constant pH 8. Fluctuations in pH have been associated with “floaters” and unclear nuclear detail. PH Blue is designed to be unaffected by carryover and is formulated to help standardize the staining process. Available in working solution or concentrate formula.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty