Would you like to log in again?
ALLWIK® Universal Absorbent Socks, Brady® SPC® Absorbents
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
107672
10810-746CS 240.97 USD InStock
10810-746
ALLWIK® Universal Absorbent Socks, Brady® SPC® Absorbents
Spill Control Materials
Use ALLWIK® all-purpose absorbents to clean up oils, coolants, solvents, and water-based fluids throughout any facility.
  • Moldable SOCs wrap around machinery and conform to uneven surfaces to prevent leaks from reaching walkways
  • Highly versatile, grey absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids
  • Gray color masks spills and stains, helping users resist the urge to pick up the absorbent before it is fully saturated
  • Made of all-purpose polypropylene, suitable for most industrial applications
Polypropylene ALLWIK® SOCs are wringable, incinerable, and suitable for most industrial applications.
Certifications: EPA 40 CFR 112.7(c)(1)(vii). OSHA 29 CFR 1910.120(j)(1)(vii). OSHA 29 CFR 1910.22(a)(2).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty