Would you like to log in again?
UltraSORB® Wipers, Foamtec International WCC
Supplier: Foamtec
UltraSORB®
HT4666 HT4699 HT4644
10810-836CS 1377.13 USD InStock
10810-836 10810-842 10810-904
UltraSORB® Wipers, Foamtec International WCC
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wipers are made from a fully cross-linked, naturally hydrophillic polyurethane foam and laundered with a proprietary process in an ISO Class 4 (FED-STD-209E Class 10/M2.5) environment.
  • UltraSORB® foam has a 3 lbs./ft³ density
  • Also offer outstanding wet tensile strength and abrasion resistance
Wipers are particle-trapping, abrasion-resistant, thermally stable, solvent-resistant, nonpyrogenic, and feature a sponge-like feel and softness, making them suitable for the wipedown of ISO Class 3 and Class 4 (FED-STD-209E Class 1/M1.5 and Class 10/M2.5) cleanrooms, optical surfaces, and hot and aseptic fill areas.
They are engineered specifically to minimize contaminants released from the wiper while maximizing the amount of particulates and residue removed from surfaces.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty