Would you like to log in again?
MRO Plus® Heavyweight Universal Absorbent Roll, Brady® SPC® Absorbents
Supplier: Brady
107732
10810-844RL 435.19 USD InStock
10810-844
MRO Plus® Heavyweight Universal Absorbent Roll, Brady® SPC® Absorbents
Spill Control Materials
MRO Plus® products are strong, durable, and low linting, making them the perfect go-to products for nearly every industrial setting.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids
  • Heavyweight design specifically targets continuous leaks and larger spills
  • Three-ply meltblown polypropylene construction is designed for general industrial applications
  • Perforations provide cost control and reduce waste by allowing users to only utilize amounts needed
The MRO Plus® Heavy Roll absorbs all industrial liquids (water, petroleum-based, and chemical-based fluids). Three-ply construction makes it stronger, more durable, and reduces lint. Perforated every 30".
Certifications: EPA 40 CFR 112.7(c)(1)(vii). OSHA 29 CFR 1910.120(j)(1)(vii). OSHA 29 CFR 1910.22(a)(2).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty