Would you like to log in again?
SWAB ULTRASOLV POINTDTIP PK500
Supplier: FOAMTEC INTERNATIONAL, LLC.
Ultrasolv* Swab^ Uniquely absorbent, with extremely low-extractable polyurethane foam heads and polypropylene handles. Streak-free, continuous Class M2.5 (Class 10) dry or low-solvent cleaning. Surfaces are cleaned without abrasion, stress, fibers, granulations, or particles. Pointed tip.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SWAB ULTRASOLV POINTDTIP PK500 10810-914 HT1750-500 Case of 12500 Retrieving
Pack of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty