Would you like to log in again?
MiraWIPE® Wipers, Foamtec International
Supplier: Foamtec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MiraWIPE®
HT5740B HT5790 HT5790B HT5790S HT5790S-25
89066-272CS 3001.98 USD InStock
89066-272 10810-940 89069-040 10810-942 10810-938
MiraWIPE® Wipers, Foamtec International
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Constructed from continuous filament microfiber polyester, woven wipers feature superior absorbency and contamination pick-up when compared to traditional knitted polyester wipers.
Woven, abrasion-resistant fabric minimizes particle generation from wiper face and edge.
Ordering information: Cleanroom laundered and packaged. Wipers are available in configurations to meet ISO Class 3 to 6 requirements; contact your VWR sales representative for more information.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty