Would you like to log in again?
UltraSOLV® Chamber Cleaning Pads, Wipers, and Swabs, Foamtec International
Supplier: Foamtec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
UltraSOLV®
HT1702-50 HT4560-10 HT4580D-10 HT4513PD-10 HT4540-10 HT4754 HT4528D-10 HT4536DS-10 HT4540D-10 HT4790-5 HT4580A-10 HT4790 HT4518D-10 HT4522-10 HT4536D-10 HT1701-5 HT4520PA-10 HT1732-50 HT4512P-10 HT1700-5 HT4510-10 HT4580-10 HT1714-50 HT4500-TXX2 HT4528-10
10811-751CS 753.12 USD InStock
10811-751 10811-773 10810-907 10811-771 10811-766 10811-787 10811-785 10811-769 10810-936 10810-952 10810-954 10810-998 89068-844 10811-783 10810-816 10810-818 10811-781 10811-777 10811-775 10811-753 10811-759 10811-758 77776-834 10811-779 10810-960
UltraSOLV® Chamber Cleaning Pads, Wipers, and Swabs, Foamtec International
Wipes Cleaning Kits and Pads
Tools are designed to work together to clean recessed surfaces and microporous sub-surfaces without using hazardous solvents. When used in conjunction, they safely and quickly remove build-up on process equipment and reduce contamination levels. Choose from diamond, silicon carbide, or aluminum oxide ScrubPADS for initial scouring. Complete the cleaning process with hydrophilic polyurethane foam sponges, wipers, and swabs.
  • Use ScrubPADS for initial scouring
  • Complete cleaning process with foam sponges, wipers, and swabs
ScrubPADS are fully laundered grits that are permanently bonded and encapsulated to stable, fabric backing.
Foam sponges, wipers, and swabs complete the cleaning of surfaces, recessed surfaces, and microporous sub-surfaces. Abrasion-resistant products absorb contamination and moisture with low particle transfer. They enable most cleaning to be done using deionized water, reducing the amount of chemistry and wiping materials required. Static-dissipative properties allow for safe cleaning of static-sensitive assemblies.
Chamber cleaning products are also offered in three convenient kits: a starter kit, customer evaluation kit, and chamber cleaning kit. Customer Evaluation Kit 10810-950 has one sample of each product and offers a convenient opportunity for reviewing the benefits and features of these chamber cleaning products with your staff. Chamber cleaning kit 89068-844 combines samples of ScrubPADS and UltraSOLV® cleaning tools.
The fabric backing is then permanently laminated to the UltraSOLV® high-absorbency foam wiping material. Diamond ScrubPADS are the most aggressive pad for removing extremely hard, glass-like build-up, such as that from titanium nitride. Silicon carbide ScrubPADS are designed for hard-surface cleaning. Pads offer long-term abrasiveness for quick removal of metal and hard polymer build-up on aluminum, stainless steel, and ceramic surfaces such as those found in implant, CVD, epitaxy, and floatzone reactors and polishers. The result is a satin-polished surface when some minor abrasion of the surface is permissible. Aluminum oxide ScrubPADS are designed for delicate surface cleaning. Use to remove hardened build-up from easily scratched tool surfaces such as anodized aluminum, quartz, polymer, and even LEXAN®. Pads gently scour and polish, leaving these surfaces clean of build-up without scratches.
Ordering information: When contacting VWR to place an order for a kit, you may customize your Starter Kit 10810-876 to include only the ScrubPAD grits needed to do one complete cleaning (all the other wiping products are included and should be used).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty