Would you like to log in again?
Accessories for Aerosol Can Recycler, New Pig
Supplier: New Pig
DRM1236 DRM1151 DRM1237 DRM1150
10811-226EA 317 USD InStock
10811-226 10811-228 10811-230 10811-232
Accessories for Aerosol Can Recycler, New Pig
Safety Cans
These replacement parts are available to keep your PIG Aerosol Can Recycler in good working order.
  • Replace the worn out neoprene gasket in your PIG Aerosol Recycler for maximum vapor control with 10811-232
  • Upgrade to a Viton gasket for extra chemical resistance in your PIG Aerosol Recycler with 10811-230
  • Replace the carbon cartridge portion of the combination filter in your PIG Aerosol Can Recycler with 10811-228
  • Replace the entire combination filter in your PIG Aerosol Can Recycler system with 10811-226
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty