Would you like to log in again?
H2O TANK WITH CAPS (UDK1X9)
Supplier: VELP SCIENTIFIC INC.
H2O tank^ H2O tank (20-liter) with caps, for Kjeldahl Distillation Unit, applications: determining ammoniacal nitrogen, protein determination, nitrogen content, nitric nitrogen (after reduction/Devarda), phenols, volatile acids, cyanides and alcohol content
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
H2O TANK WITH CAPS (UDK1X9) 10811-386 A00000266 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty