Would you like to log in again?
TUBE CRYO 2D CD CLAS VI MED-GRD HDPE 5ML
Supplier: Thermo Fisher Scientific
Tube^ Cryo tube with 2D barcode, Closure Material: Virgin Class VI Medical-Grade HDPE with HDPE Gasket Material, Autoclavable, Universal cap, DNase Free, External thread, SAL of 10-6, Sterile, White patch and Graduations, RNase Free, Volume: 5ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE CRYO 2D CD CLAS VI MED-GRD HDPE 5ML 10811-404 374320-BR Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty