Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® 12" Test Sieves, Intermediate Height, Brass and Stainless Steel
Supplier: VWR
18BS12I 1/4BS12I 3.5BS12I 140BS12I 16BS12I 50BS12I 7/16BS12I 100BS12I 20BS12I 4BS12I 1/8BS12I 60BS12I 1/2BS12I 5/16BS12I 30BS12I 230BS12I 35BS12I 500BS12I 5/8"BS12I 200BS12I 635BS12I 6BS12I 5BS12I 170BS12I 270BS12I 80BS12I 120BS12I 1/4_BS12I 400BS12I 70BS12I 40BS12I 12BS12I .265BS12I 25BS12I 8BS12I 7BS12I 325BS12I 450BS12I 45BS12I 3/8BS12I 10BS12I
47730-038EA 327.66 USD InStock
47730-038 47730-112 47730-058 47730-078 10811-590 10811-630 47730-098 10811-530 47730-096 10811-722 10811-622 47730-094 10811-720 47730-092 47730-070 10811-620 10811-528 47730-090 10811-626 10811-724 10811-624 10811-628 10811-808 47730-106 47730-104 47730-068 47730-102 47730-066 10811-784 47730-086 47730-084 47730-082 10811-710 10811-534 10811-632 10811-532 10811-714 10811-712 10811-718 10811-618 10811-716
VWR® 12" Test Sieves, Intermediate Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Intermediate Height (2") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty