Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® 12" Test Sieves, Half Height, All Stainless Steel
Supplier: Avantor
8SS12H 120SS12H .530SS12H 40SS12H 5/8SS12H .265SS12H 14SS12H 20SS12H 7/16SS12H 170SS12H 25SS12H 6SS12H 1/4SS12H 5/16SS12H 4SS12H 325SS12H 80SS12H 35SS12H 30SS12H 500SS12H 200SS12H 270SS12H 400SS12H 70SS12H 635SS12H 45SS12H 230SS12H 100SS12H 140SS12H 18SS12H 3.5SS12H 1/2SS12H 12SS12H 60SS12H 7SS12H 450SS12H 1-1/2SS12H 5SS12H 7/8SS12H 3/8"SS12H 1.00SS12H 10SS12H 1/8SS12H 50SS12H 16SS12H 3/4SS12H
47730-318EA 456.03 USD InStock
47730-318 47730-314 47730-310 10811-570 47730-330 10811-652 10811-574 47730-274 10811-552 10811-650 10811-572 10804-776 10811-550 10811-568 10811-644 10811-566 10811-588 10811-664 10811-642 10811-648 10811-646 47730-306 47730-346 10811-560 10811-582 47730-300 10811-580 10811-564 10811-586 10811-640 10811-662 10811-584 10811-562 10811-660 10811-656 10811-634 10811-578 10811-556 10811-654 10811-576 10811-554 10811-638 10811-636 10811-658 10811-558 10804-790
VWR® 12" Test Sieves, Half Height, All Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm ) all stainless steel test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Half Height (15/8") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying the sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes, and Centerline® sieve inspection or calibration certificates are available as accessories.
Certifications: Each serialized sieves comes with a dated ASTM E11 Certificate of Compliance.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth. Sieve Certification, Pans, and Covers must be ordered separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty