Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 12" Test Sieves, Intermediate Height, Brass and Stainless Steel

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

18BS12I 1/4BS12I 3.5BS12I 140BS12I 16BS12I 50BS12I 7/16BS12I 100BS12I 20BS12I 4BS12I 1/8BS12I 60BS12I 1/2BS12I 5/16BS12I 30BS12I 230BS12I 35BS12I 500BS12I 5/8"BS12I 200BS12I 635BS12I 6BS12I 5BS12I 170BS12I 270BS12I 80BS12I 120BS12I 1/4_BS12I 400BS12I 70BS12I 40BS12I 12BS12I .265BS12I 25BS12I 8BS12I 7BS12I 325BS12I 450BS12I 45BS12I 3/8BS12I 10BS12I
47730-038EA 327.66 USD InStock
47730-038 47730-112 47730-058 47730-078 10811-590 10811-630 47730-098 10811-530 47730-096 10811-722 10811-622 47730-094 10811-720 47730-092 47730-070 10811-620 10811-528 47730-090 10811-626 10811-724 10811-624 10811-628 10811-808 47730-106 47730-104 47730-068 47730-102 47730-066 10811-784 47730-086 47730-084 47730-082 10811-710 10811-534 10811-632 10811-532 10811-714 10811-712 10811-718 10811-618 10811-716
VWR® 12" Test Sieves, Intermediate Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.

  • Intermediate Height (2") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction

Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.

Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.

Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty