Would you like to log in again?
HydroGOLD Capillary Columns
Supplier: Thermo Scientific
HydroGOLD
26H50-0525 26H50-1032 26H50-0532 26H50-0553 26H50-0253 26H50-0232 26H50-1053
10811-886EA 101.8 USD InStock
10811-886 10811-884 10811-902 10811-900 10811-890 10811-894 10811-892
HydroGOLD Capillary Columns
Chromatography Columns
Capillary HydroGOLD columns provide protection to the analytical column.
  • Focuses target analytes at the head of the analytical column, leading to better chromatographic peak shape
  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • HydroGOLD extends column lifetime by preventing degradation from harsh 'steam-cleaning' water injections
Protects against column contamination caused by non-volatile materials, extending the column lifetime.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty