Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples.
Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1 mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150000 plates per meter for the 3 µm particle size and 100000 plates per meter for the 5 µm particle size, Purospher™ STAR columns ensure good baseline separations. Special derivatization and endcapping processes ensure long-term chemical stability, excellent retention stability, and long column lifetime. All Purospher™ STAR columns are guaranteed to provide absolutely reproducible results.
Ordering information: All LiChroCART® cartridges require end fittings. manuCART® and LiChroCART® series end fittings and holders are available separately. Hibar® columns include end fittings. If 4×4 mm guard columns are to be used on 4 or 4.6 mm Int.Ø. Hibar® Purospher™ STAR columns, the top fitting must be replaced with guard column holder designed to accommodate this size guard column. Other larger Int.Ø. Purospher™ STAR columns include end fittings as indicated in their descriptions. Guard columns for these larger Int.Ø columns are also available, contact your Avantor sales representative to order.