Would you like to log in again?
Hettich ROTINA 380 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Supplier: Hettich
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1706-01 380RCELLCUL4BC 380CELLCULT4 380BLOOD4 380BOTTLE 380RCELLCUL2BC 380BLOOD2 380RBLOOD4-BC 380RCELLCULT2 380RBOTTLE 380BOTTLE-BC 380BLOOD4-BC 380CELLCULT4BC 380CELLCULT2 380RBLOOD2 380CELLCULT2BC 380RCELLCULT4 380PLATE 380RPLATE 380RBLOOD4 380RBLOOD2-BC 380RBOTTLE-BC 1701-01 380BLOOD2-BC
10812-378EA 14225.94 USD InStock
10812-378 10812-268 10812-388 10812-256 10812-258 10813-206 75801-818 10812-390 10812-392 10812-260 10812-380 10812-270 10812-262 10812-394 10812-372 75801-820 10812-382 10812-374 10812-264 10812-384 10812-266 10812-376 10812-386 10812-254
Hettich ROTINA 380 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
The Hettich Lab Technology ROTINA 380 benchtop centrifuge series delivers versatility, capacity and performance in a compact footprint.
  • Max capacity – 4 x 290 mL
  • Max RPM/RCF – 15,000/24,400
  • Temperature control available
  • General purpose
  • Versa-Rotor (1 rotor for multiple applications)
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design - easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety - self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty - all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904
Switch applications without switching rotors with the Hettich Versa-Rotor
Convenient application packages for blood tubes, cell cultures, plates, and bottles are available. Includes everything you need to perform your application: centrifuge, rotor, buckets, and lids, where applicable.
Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.
Ideal for mid-sized clinical, research and industrial applications. Maximum capacity of 4 x 290 mL, 76 blood tubes, 36 x 15/50 mL conical tubes, or 12 microtiter plates. Robust temperature controlled model (ROTINA 380 R) available for temperature-sensitive samples.
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Hettich Lab Technology centrifuge packages include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable. Supports over 234 application configurations; please contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty