Would you like to log in again?
Hettich EBA 200 Clinical Tabletop Centrifuges, Hettich Lab Technology
Supplier: Hettich
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
200URINALYSIS4 200BLOOD8 1802-01 200BLOOD8-PED 1800-01 200SBLOOD8 200SBLOOD8-PED
10812-620EA 2568.91 USD InStock
10812-620 10812-612 75801-628 75801-626 10812-614 10812-616 10812-618
Hettich EBA 200 Clinical Tabletop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
Hettich Lab Technology's EBA 200 tabletop clinical centrifuge series offers powerful performance and easy operation in a compact footprint.
  • STAT capable (EBA 200 S)
  • Max RPM/RCF – 8,000/6,153
  • Built-in 8-place rotor
  • Compact and powerful for standard blood, urine, and pediatric tubes
  • All-metal containment
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design – easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety – self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty – all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904
The EBA 200 is ideal for small-batch clinical chemistry applications, and the higher-performance EBA 200 S model is STAT capable, achieving PPP in minutes
Convenient application packages for blood tubes, pediatric tubes, and urinalysis are available. Includes everything you need to perform your application: centrifuge, rotor, and adapters.
Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.
Includes a built-in 8-place rotor.
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Hettich Lab Technology centrifuge packages include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty