Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hettich EBA 280 Clinical Tabletop Centrifuges, Hettich Lab Technology

Supplier: Hettich

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

280BLOOD6 280BLOOD6-PED 280BLOOD8 280SEROLOGY-D 280BLOOD12 1101-01 280SBLOOD6 280SEROLOGY 280URINALYSIS6 1102-01
10813-132EA 6160.53 USD InStock
10813-132 75802-818 10813-130 10812-624 10812-626 75802-820 10813-128 10813-126 10813-124 10813-134
Hettich EBA 280 Clinical Tabletop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
The Hettich Lab Technology EBA 280 clinical tabletop centrifuge series delivers 90° flat separation for all standard blood, pediatric and urine (collection and analysis) tubes.

  • 90° flat separation
  • STAT (EBA 280 S)
  • Quick-Lift rotor
  • Serology centrifuge
  • Max capacity – 6 x 50 mL
  • Max RPM/RCF – 6,000/5,071
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design – easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety – self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty – all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – Engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904

The higher-performance EBA 280 S model is STAT capable, delivering PPP in minutes

Convenient application packages for blood tubes, STAT, serology, serology decanting, pediatric tubes, urinalysis, and cell cultures are available. Includes everything you need to perform your application: centrifuge, rotor, buckets, and adapters.

Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.

Supports the Hettich Quick-Lift rotor for effortless rotor installation (no tools required) and sample management.

Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty