Would you like to log in again?
Hettich ROTINA 420 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Supplier: Hettich
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4706-01 420CELLCULT2BC 420BLOOD2-BC 420RCELLCUL4BC 420BLOOD4-BC 420RBOTTL400BC 420RBLOOD2-BC 420RBOTTL250BC 4701-01 420RBLOOD4-BC 420BOTTLE400BC 420PLATE 420BOTTLE250BC 420CELLCULT4BC 420RCELLCUL2BC 420RPLATE
10812-278EA 25410.88 USD InStock
10812-278 10813-158 10813-156 75802-372 10813-154 75802-370 10812-280 10812-290 10812-282 10812-284 10812-272 10812-396 10812-286 10812-274 10812-288 10812-276
Hettich ROTINA 420 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
Hettich Lab Technology's ROTINA 420 centrifuge series delivers high capacities and versatility right on the benchtop.
  • Max capacity – 4 x 600 mL
  • Max RPM/RCF – 15,000/24,400
  • General-purpose
  • Versa-Rotor (1 rotor for multiple applications)
  • Temperature controlled available
  • Large volumes
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design– easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety– self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty– all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904
Switch applications without switching rotors with the Hettich Versa-Rotor
Convenient application packages for blood tubes, cell cultures, plates, and bottles are available. Includes everything you need to perform your application: centrifuge, rotor, buckets, and lids, where applicable.
Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.
Maximum capacity of 4 x 600 mL, 140 blood tubes, 52 x 15 mL conical tubes, or 16 microtiter plates. Robust temperature controlled model available (ROTINA 420 R) for temperature-sensitive samples.
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Hettich Lab Technology centrifuge packages include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable. Supports over 302 application configurations; please contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty