Would you like to log in again?
Hettich ROTANTA 460 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Supplier: Hettich
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
460RBLOOD4B-BC 460RPLATEHS-BC 460CELLCULTHC 460BLOODHC-BC 460RBLOODHC-BC 460RCELCULHCBC 460RBLDBAG4-BC 460CELLCULHCBC 460BOTTLE250BC 460RCELLCUL2BC 460BLOODHC 460RPLATE-BC 460CELLCULT2BC 460BOTTLE750BC 460RBOTTL250BC 460RBLOODBAG4 460RBLOODHC 460RCELLCUL4BC 460RBLOOD2-BC 460PLATE-BC 460BLOOD4B-BC 460CELLCULT4BC 460RBOTTL750BC 460PLATE 460RPLATE 460PLATEHS-BC 5650-01 5660-01 460RBLOOD4-BC 460BLOOD4-BC 460RCELLCULTHC 460BLOOD2-BC
10812-312EA 25349.49 USD InStock
10812-312 10812-314 10812-316 10812-318 75802-452 10813-164 10813-162 10813-160 10812-420 10812-320 10812-422 10812-400 10812-300 10812-322 10812-424 10812-402 10812-302 10812-324 10812-326 10812-304 10812-306 10812-328 10812-308 75802-450 10812-292 10812-294 10812-330 10812-296 10812-310 10812-398 10812-332 10812-298
Hettich ROTANTA 460 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
The Hettich Lab Technology ROTANTA 460 benchtop centrifuge series processes large sample volumes at high g-forces for high-throughput laboratories.
  • Max capacity – 4 x 750 mL/3 liters
  • Max RPM/RCF – 15,000/24,400
  • High throughputs
  • Specialty applications
  • Versa-Rotor (1 rotor for multiple applications)
  • Temperature control available
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design – easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety – self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty – all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904
Switch applications without switching rotors with the Hettich Versa-Rotor
Convenient application packages for blood tubes, blood bags, cell cultures, plates, and bottles are available. Includes everything you need to perform your application: centrifuge, rotor, buckets, and lids, where applicable.
Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.
Supports a variety of accessories for specialty applications such as a high-speed plate rotor and accessories for flasks, chrome bath and schlenk tubes. Maximum capacity of 4 x 750 mL, 180 blood or urine tubes, 96 x 15 mL, 40 x 50 mL conical tubes, 24 microtiter plates, and more. Robust temperature control available (ROTANTA 460 R) for temperature-sensitive samples.
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Hettich Lab Technology centrifuge packages include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable. Supports over 470 application configurations; please contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty