Would you like to log in again?
Flame Resistant Denim Work Jeans with Max Comfort™ Material, MCR Safety
Supplier: MCR Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
P1D5034 P1D4636 P1D4632 P1D3430 P1D3830 P1D3436 P1D2834 P1D4030 P1D4034 P1D4036 P1D4230 P1D4236 P1D5032 P1D3836 P1D4430 P1D2830 P1D2832 P1D3832 P1D3632 P1D3236 P1D4234 P1D4832 P1D4434 P1D3434 P1D4634 P1D3034 P1D3030 P1D3630 P1D3634 P1D3432 P1D3230 P1D4836 P1D3234 P1D3834 P1D3036 P1D4032 P1D5030 P1D5036 P1D4630 P1D4830 P1D4232 P1D4834 P1D3636 P1D3032 P1D4432 P1D4436 P1D3232
10813-318EA 106.7 USD InStock
10813-318 10813-338 10813-316 10813-358 10813-336 10813-314 10813-560 10813-356 10813-334 10813-378 10813-312 10813-332 10813-562 10813-354 10813-376 10813-310 10813-386 10813-342 10813-364 10813-320 10813-384 10813-362 10813-340 10813-382 10813-360 10813-380 10813-558 10813-308 10813-306 10813-328 10813-326 10813-348 10813-304 10813-368 10813-324 10813-346 10813-344 10813-322 10813-388 10813-366 10813-564 10813-330 10813-374 10813-352 10813-372 10813-350 10813-370
Flame Resistant Denim Work Jeans with Max Comfort™ Material, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics.
  • Enzyme wash for softness, comfort, and look
  • 8″ single needle topstitch at top of outseam for durability
  • 100% cotton
  • NFPA 2112, NFPA 70E, UL certified
The P1 Work Jeans feature a comfortable 100% cotton material, and meet the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL certified.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty