Would you like to log in again?
Flame Resistant Work Shirt with Max Comfort™ Material, Tan, MCR Safety
Supplier: MCR Safety
S1TLT S1TX3T S1TL S1TMT S1TX4T S1TX2T S1TX4 S1TM S1TX2 S1TX3 S1TXL S1TXLT
10813-442EA 141.84 USD InStock
10813-442 10813-432 10813-444 10813-434 10813-446 10813-436 10813-448 10813-438 10813-450 10813-440 10813-430 10813-452
Flame Resistant Work Shirt with Max Comfort™ Material, Tan, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
  • Comfortable 88% cotton/12% nylon blend
  • Two chest pockets
  • NFPA 2112, NFPA 70E, UL Certified
The S1 Work Shirt features a comfortable 88% cotton/12% nylon blend that meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty