Would you like to log in again?
Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Tan, MCR Safety
Supplier: MCR Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Max Comfort™
H1TX3 H1TX4 H1TXL H1TL H1TS H1TX5 H1TM H1TX2
10813-486EA 135.38 USD InStock
10813-486 10813-498 10813-488 10813-500 10813-490 10813-492 10813-494 10813-496
Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Tan, MCR Safety
Workwear
The H1 Henley Shirt features a 100% FR Cotton material for Maximum Comfort. The Max Comfort™ material meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace.
  • Left chest pocket
  • Moisture wicking and breathable
  • Rib-knit collar and cuff
All MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty