Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloropurine 2'-deoxyriboside 97%

Supplier: Thermo Scientific Chemicals
Warning

J64871-03 J64871-03 J64871-MD
10814-350EA 98.92 USD InStock
10814-350 AAJ64871-03 CAAAJ64871-03
6-Chloropurine 2'-deoxyriboside 97%
6-Chloropurine 2'-deoxyriboside
Powder


Formula: C₁₀H₁₁ClN₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00083282
CAS Number: 4594-45-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty