Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,3,5-Tetra-O-acetyl-β-L-ribofuranose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3S,4S,5S)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

J66074-06 J66074-09 J66074-03
10814-408EA 43.92 USD InStock
10814-408 10814-412 10814-410
1,2,3,5-Tetra-O-acetyl-β-L-ribofuranose 98%
1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose


Formula: C₁₃H₁₈O₉
MW: 318.278 g/mol
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00674547
CAS Number: 144490-03-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty