Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylumbelliferyl phosphate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methylumbelliferyl phosphoric acid4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphatePhosphoric Acid 4-Methylumbelliferyl Ester

J66091-MD J66091-03 J66091-06
10814-428EA 528.44 USD InStock
10814-428 10814-426 10814-430
4-Methylumbelliferyl phosphate 99%
4-Methylumbelliferyl phosphate


Formula: C₁₀H₉O₆P
MW: 256.15 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Freezer
MDL Number: MFCD00016969
CAS Number: 3368-04-5
EINECS: 222-137-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty