Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylumbelliferyl sulfate potassium salt 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Potassium 4-methyl-2-oxo-2H-chromen-7-yl sulphatePotassium 4-methyl-2-oxo-2H-chromen-7-yl sulfate

J66160-ME J66160-03
10814-460EA 241.37 USD InStock
10814-460 10814-464
4-Methylumbelliferyl sulfate potassium salt 98%
4-Methylumbelliferyl sulfate potassium salt
Substrate for sulfatase


Formula: C₁₀H₇KO₆S
MW: 294.318 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00016970
CAS Number: 15220-11-8
EINECS: 239-272-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty