Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Fructose-1,6-bisphosphate trisodium salt octahydrate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium (3S,4R,5R)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate octahydrate

J66256-14 J66256-22 J66256-09
10814-538EA 128.84 USD InStock
10814-538 10814-536 10814-540
D-Fructose-1,6-bisphosphate trisodium salt octahydrate 98%
D-Fructose-1,6-bisphosphate trisodium salt octahydrate


Formula: C₆H₂₇Na₃O₂₀P₂
MW: 550.179 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00149340
CAS Number: 81028-91-3
EINECS: 253-778-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty