Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Glucosamine sulfate 98%

Supplier: Thermo Scientific Chemicals

J66271-14 J66271-06
10814-554EA 46.74 USD InStock
10814-554 10815-476
D-Glucosamine sulfate 98%
D-Glucosamine sulfate


Formula: C₆H₁₅NO₉S
MW: 277.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080790
CAS Number: 29031-19-4
EINECS: 249-379-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty