Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D-Galactose pentaacetate

Supplier: Thermo Scientific Chemicals
Synonyms: a-D-Galactose pentaacetate

J66366-14 J66366-06 J66366-18
10814-582EA 283.77 USD InStock
10814-582 10814-580 10814-578
α-D-Galactose pentaacetate
alpha-D-Galactose pentaacetate


Formula: C₁₆H₂₂O₁₁
MW: 390.341 g/mol
Melting Pt: 143…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00064081
CAS Number: 4163-59-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty