Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decyl-β-D-glucopyranoside

Supplier: Thermo Scientific Chemicals
Synonyms: Decyl glucopyranosideDGN-Decyl beta-D-glucopyranosideDecyl beta-D-glucopyranoside

J66423-03 J66423-06
10814-632EA 539.2 USD InStock
10814-632 10814-630
Decyl-β-D-glucopyranoside
Decyl β-D-glucopyranoside
Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.


Formula: C₁₆H₃₂O₆
MW: 320.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063297
CAS Number: 58846-77-8
EINECS: 261-469-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty