Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Fructose-16-diphosphate dicalcium salt 95%

Supplier: Thermo Scientific Chemicals

J66477-30 J66477-22 J66477-14
10815-578EA 331.75 USD InStock
10815-578 10814-664 10814-662
D-Fructose-16-diphosphate dicalcium salt 95%
D-Fructose-16-diphosphate dicalcium salt


Formula: C₆H₁₀Ca₂O₁₂P₂
Storage Temperature: Ambient
MDL Number: MFCD00135875
CAS Number: 6055-82-9
EINECS: 227-979-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty