Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Hexadecyl-β-D-maltoside 98%

Supplier: Thermo Scientific Chemicals
Synonyms: n-Hexadecyl β-D-maltopyranoside(2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Hexadecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

J66531-ME J66531-MC
10814-682EA 134.34 USD InStock
10814-682 10814-684
n-Hexadecyl-β-D-maltoside 98%
n-Hexadecyl-β-D-maltoside


Formula: C₂₈H₅₄O₁₁
MW: 566.72 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01310880
CAS Number: 98064-96-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty