Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7)

Supplier: Thermo Scientific
J66592-MB J66592-MA
10814-728EA 287.09 USD InStock
10814-728 10814-730
[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7)
Proteins and Peptides
Molecular Formula: C39H59N11O9S
Formula Weight: 858.02
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty