Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-O-ɑ-D-glucopyranosyl-L-ascorbic acid 99.5%

Supplier: Thermo Scientific Chemicals
Synonyms: AA-2G

J66601-03 J66601-06
10814-738EA 113.89 USD InStock
10814-738 10814-736
2-O-ɑ-D-glucopyranosyl-L-ascorbic acid 99.5%
2-O-alpha-D-glucopyranosyl-L-ascorbic acid
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduces the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. 2-O-alpha-D-Glucopyranosyl-L-ascorbic acid also promotes collagen synthesis and function as an antioxidant.


Formula: C₁₂H₁₈O₁₁
MW: 338.27 g/mol
Melting Pt: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00925623
CAS Number: 129499-78-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty