Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3:4,5-Di-O-isopropylidene-D-arabitol 98%

Supplier: Thermo Scientific Chemicals

J66608-03 J66608-ME
10814-742EA 86.45 USD InStock
10814-742 10814-740
2,3:4,5-Di-O-isopropylidene-D-arabitol 98%
2,3:4,5-Di-O-isopropylidene-D-arabitol


Formula: C₁₁H₂₀O₅
Storage Temperature: Freezer
MDL Number: MFCD00063387
CAS Number: 19139-74-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty