Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetatea-D-Mannose pentaacetate

J66685-14 J66685-06
10814-758EA 88.25 USD InStock
10814-758 10814-760
1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose 98%
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose


Formula: C₁₆H₂₂O₁₁
MW: 390.341 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00069798
CAS Number: 4163-65-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty