Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylphenyl-β-D-thiogalactopyranoside 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methylphenyl 1-thio-beta-D-galactopyranoside

J66686-06 J66686-03
10814-764EA 251.89 USD InStock
10814-764 10814-762
4-Methylphenyl-β-D-thiogalactopyranoside 98%
4-Methylphenyl-beta-D-thiogalactopyranoside


Formula: C₁₃H₁₈O₅S
MW: 286.342 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973631
CAS Number: 28244-98-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty