Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sucralose 98%, powder

Supplier: Thermo Scientific Chemicals
Synonyms: 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

J66736-30 J66736-22 J66736-18
10814-816EA 267.73 USD InStock
10814-816 10814-814 10814-812
Sucralose 98%, powder
Sucralose
A synthetic sweetener approximately 600 times sweeter than sucrose. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells


Formula: C₁₂H₁₉Cl₃O₈
MW: 397.63 g/mol
Melting Pt: 130 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03648615
CAS Number: 56038-13-2
EINECS: 259-952-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty