Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cecropin A (2-9) Peptide

Supplier: Thermo Scientific
J66741-MCR J66741-EXD
10814-822EA 230.3 USD InStock
10814-822 10814-820
Cecropin A (2-9) Peptide
Proteins and Peptides
Molecular Formula: C89H152N22O15
Formula Weight: 1770.29
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD01630779

Causes the release of carboxyfluoresceine encapsulated in phosphatidylcholine liposomes.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty