Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caspase-3-substrate III

Supplier: Thermo Scientific Chemicals

J66881-EXD J66881-MCR
10814-888EA 301.3 USD InStock
10814-888 10814-886
Caspase-3-substrate III
Caspase-3-substrate III


Formula: C₅₀H₆₂N₁₀O₂₂
MW: 1155.094 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798785
CAS Number: 189696-20-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty