Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tri-O-acetyl-D-galactal

Supplier: Thermo Scientific Chemicals
Synonyms: 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol 3,4,6-Tri-O-acetyl Ether

J66904-06 J66904-09 J66904-03
10814-902EA 253.15 USD InStock
10814-902 10814-900 10814-898
Tri-O-acetyl-D-galactal
Tri-O-acetyl-D-galactal


Formula: C₁₂H₁₆O₇
MW: 272.25 g/mol
Melting Pt: 34…38 °C
Flash Pt: >110 °C(235 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00064092
CAS Number: 4098-06-0
EINECS: 223-859-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty