Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Keto-D-gluconic acid potassium salt 98%

Supplier: Thermo Scientific Chemicals
Warning

J66916-03 J66916-06
10814-916EA 273.92 USD InStock
10814-916 10814-914
5-Keto-D-gluconic acid potassium salt 98%
5-Keto-D-gluconic acid potassium salt


Formula: C₆H₉KO₇
Storage Temperature: Refrigerator
MDL Number: MFCD00069562
CAS Number: 5447-60-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty