Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Trehalose, anhydrous, powder

Supplier: Thermo Scientific Chemicals
Synonyms: (+)-TrehaloseTrehaloseD-Trehaloseα-D-Glucopyranosyl-α-D-glucopyranosideα,α-Trehalose

J66006-18 J66006-09 J66006-06
10815-360EA 66.01 USD InStock
10815-360 10815-362 10815-358
D-(+)-Trehalose, anhydrous, powder
D(+)-Trehalose


Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006628
CAS Number: 99-20-7
EINECS: 202-739-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty